Merck
  • Hawthorn ethanolic extracts with triterpenoids and flavonoids exert hepatoprotective effects and suppress the hypercholesterolemia-induced oxidative stress in rats.
Hawthorn ethanolic extracts with triterpenoids and flavonoids exert hepatoprotective effects and suppress the hypercholesterolemia-induced oxidative stress in rats.
Iranian journal of basic medical sciences (2015-09-12)
Ali Rezaei-Golmisheh, Hassan Malekinejad, Siamak Asri-Rezaei, Amir Abbas Farshid, Peyman Akbari
ZUSAMMENFASSUNG
The current study was aimed to determine the bioactive constituents and biological effects of the Crataegus monogyna ethanolic extracts from bark, leaves and berries on hypercholesterolemia. Oleanolic acid, ursolic acid, quercetin and lupeol concentrations were quantified by HPLC. Total phenol content and radical scavenging activity of extracts were also measured. The hypocholesterolemic, antioxidant, and hepatoprotective effects of the extracts were examined in hypercholesterolemic rats and compared with orlistat. The highest phenol content, oleanolic acid, quercetin and lupeol levels and free radical scavenging potency were found in the bark extract, and the highest ursolic acid level was found in the berries extract. Orlistat and extracts significantly (P<0.05) lowered the hypercholesterolemia-increased serum level of hepatic enzymes and lipid peroxidation level. Hawthorn's extracts protected from hepatic thiol depletion and improved the lipid profile and hepatic damages. Data suggested that hawthorn's extracts are able to protect from hypercholesterolemia-induced oxidative stress and hepatic injuries. Moreover, the hypocholesterolemic effect of extracts was found comparable to orlistat.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoesäure), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Gallussäure, 97.5-102.5% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoesäure), ReagentPlus®, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Butyliertes Hydroxytoluol, ≥99%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Thiobarbitursäure, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Ursolsäure, ≥90%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Oleanolsäure, ≥97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphorsäure, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphorsäure, BioReagent, suitable for insect cell culture, 85%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Butylalkohol, natural, ≥99.5%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Lupeol, ≥94%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phosphorsäure -Lösung, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.