Merck
Characterization of an L-amino acid oxidase in equine spermatozoa.
Biology of reproduction (2015-03-06)
Joanna B Aitken, Nenad Naumovski, Ben Curry, Christopher G Grupen, Zamira Gibb, R John Aitken
ZUSAMMENFASSUNG
This study demonstrates for the first time the presence of an L-amino acid oxidase (LAAO) enzyme in equine spermatozoa that is able to generate significant amounts of reactive oxygen species (ROS) and create a state of oxidative stress. RT-PCR analysis revealed that the mRNA for this enzyme was present in the equine testis and spermatozoa, while immunocytochemical studies demonstrated that the mature LAAO protein was located in the sperm head, particularly in the acrosomal and postacrosomal domains. Experimental studies demonstrated that the aromatic amino acids (L-phenylalanine > L-tryptophan > L-tyrosine) were substrates for this enzyme, eliciting the dose- and time-dependent generation of ROS via mechanisms that were enhanced by cell death. This unexpected result was confirmed by analyses of ROS generation in subcellular sperm fractions, which again located a majority of LAAO activity to the sperm head. Equine cryopreservation medium was shown to contain sufficient quantities of aromatic amino acids to activate the LAAO system and generate ROS. The biological significance of this activity was established in an experiment in which physiological concentrations of aromatic amino acids were found to suppress sperm motility but only if dead spermatozoa were present in the same suspension. The combination of aromatic amino acids and nonviable cells was also found to enhance the levels of lipid peroxidation in live spermatozoa. These results suggest the potential significance of LAAO activity in generating the oxidative stress associated with the cryopreservation of equine spermatozoa. It is possible that inhibitors of this enzyme system may facilitate the development of modified cryostorage regimes for clinical validation in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Azodicarbonsäure-diethylester -Lösung, purum, ~40% in toluene (H-NMR)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, tablet
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Preise und Verfügbarkeit sind derzeit nicht verfügbar.