Merck
  • Charge-controlled nanoprecipitation as a modular approach to ultrasmall polymer nanocarriers: making bright and stable nanoparticles.
Charge-controlled nanoprecipitation as a modular approach to ultrasmall polymer nanocarriers: making bright and stable nanoparticles.
ACS nano (2015-04-22)
Andreas Reisch, Anne Runser, Youri Arntz, Yves Mély, Andrey S Klymchenko
ZUSAMMENFASSUNG
Ultrasmall polymer nanoparticles are rapidly gaining importance as nanocarriers for drugs and contrast agents. Here, a straightforward modular approach to efficiently loaded and stable sub-20-nm polymer particles is developed. In order to obtain ultrasmall polymer nanoparticles, we investigated the influence of one to two charged groups per polymer chain on the size of particles obtained by nanoprecipitation. Negatively charged carboxylate and sulfonate or positively charged trimethylammonium groups were introduced into the polymers poly(d,l-lactide-co-glycolide) (PLGA), polycaprolactone (PCL), and poly(methyl methacrylate) (PMMA). According to dynamic light scattering, atomic force and electron microscopy, the presence of one to two charged groups per polymer chain can strongly reduce the size of polymer nanoparticles made by nanoprecipitation. The particle size can be further decreased to less than 15 nm by decreasing the concentration of polymer in the solvent used for nanoprecipitation. We then show that even very small nanocarriers of 15 nm size preserve the capacity to encapsulate large amounts of ionic dyes with bulky counterions at efficiencies >90%, which generates polymer nanoparticles 10-fold brighter than quantum dots of the same size. Postmodification of their surface with the PEG containing amphiphiles Tween 80 and pluronic F-127 led to particles that were stable under physiological conditions and in the presence of 10% fetal bovine serum. This modular route could become a general method for the preparation of ultrasmall polymer nanoparticles as nanocarriers of contrast agents and drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
N,N-Diisopropylethylamin, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Taurin, ≥99%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Methacrylsäure, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Natriumphosphat, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Rhodamin B, ≥95% (HPLC)
Sigma-Aldrich
N,N-Diisopropylethylamin, purified by redistillation, 99.5%
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
1-Hydroxybenzotriazol Hydrat, ≥97.0% dry basis (T)
Sigma-Aldrich
Rhodamin B, suitable for fluorescence
Sigma-Aldrich
Natriumphosphat, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Natriumphosphat, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Natriumphosphat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumphosphat, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Bernsteinsäureanhydrid, ≥99% (GC)
Sigma-Aldrich
Taurin, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
1-Hydroxybenzotriazol Hydrat, wetted with not less than 14 wt. % water, 98% dry basis
Sigma-Aldrich
Natriumphosphat, zweibasisch -Lösung, BioUltra, 0.5 M in H2O