Merck
Protective Effects of BDNF against C-Reactive Protein-Induced Inflammation in Women.
Mediators of inflammation (2015-07-15)
Nicole Noren Hooten, Ngozi Ejiogu, Alan B Zonderman, Michele K Evans
ZUSAMMENFASSUNG
Since high sensitivity C-reactive protein (hsCRP) is predictive of cardiovascular events, it is important to examine the relationship between hsCRP and other inflammatory and oxidative stress markers linked to cardiovascular disease (CVD) etiology. Previously, we reported that hsCRP induces the oxidative stress adduct 8-oxo-7,8-dihydro-2'deoxyguanosine (8-oxodG) and that these markers are significantly associated in women. Recent data indicates that brain-derived neurotrophic factor (BDNF) may have a role in CVD. We examined BDNF levels in 3 groups of women that were age- and race-matched with low (<3 mg/L), mid (>3-20 mg/L), and high (>20 mg/L) hsCRP (n = 39 per group) and found a significant association between hsCRP, BDNF, and 8-oxodG. In African American females with high hsCRP, increases in BDNF were associated with decreased serum 8-oxodG. This was not the case in white women where high hsCRP was associated with high levels of BDNF and high levels of 8-oxodG. BDNF treatment of cells reduced CRP levels and inhibited CRP-induced DNA damage. We discovered an important relationship between hsCRP, 8-oxodG, and BDNF in women at hsCRP levels >3 mg/L. These data suggest that BDNF may have a protective role in counteracting the inflammatory effects of hsCRP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
SAFC
L-Glutamin
Sigma-Aldrich
Wachstumsfaktor BDNF human, BDNF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Menadion, crystalline
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
BDNF human, Carrier free, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), suitable for cell culture
Sigma-Aldrich
Menadion, meets USP testing specifications
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications