Merck
Novel 4-heteroaryl-antipyrines as DPP-IV inhibitors.
Chemical biology & drug design (2015-06-03)
Sobhi M Gomha, Taha M A Eldebss, Mohamed G Badrey, Mohamed M Abdulla, Abdelrahman S Mayhoub
ZUSAMMENFASSUNG
Type 2 diabetes mellitus is a vast growing progressive disease that almost affects one person among every twelve globally. Regardless the availability of wide variety of oral hypoglycemics, only one-third of patients achieves proper glycemic control. With the advantage of the low risk of hypoglycemia, DPP-IV attracted the attention of medicinal chemists as a new target for oral hypoglycemics. In this report, a lead compound 1, with antipyrine scaffold, was obtained, and its binding mode was calculated. Several derivatives with bridged nitrogenous heterocycles have been synthesized via multicomponent reaction under controlled microwave heating conditions. The antidiabetic activity versus DPP-IV protein was evaluated and compared with sitagliptin. Compounds with smaller- or medium-sized nitrogenous bridges were comparable with sitagliptin in terms of DPP-IV inhibitory activity, potentially via targeting Glu203 and Glu204. The oral hypoglycemic activities of compounds with submicromolar IC50 values were further evaluated using diabetic mouse model.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Hydrazin Monohydrat, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazin Hydrat, reagent grade, N2H4 50-60 %
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Phenylhydrazin, 97%
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Hydrazinhydrat -Lösung, puriss. p.a., 24-26% in H2O (RT)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis