Merck
  • Impact of chrysosplenetin on the pharmacokinetics and anti-malarial efficacy of artemisinin against Plasmodium berghei as well as in vitro CYP450 enzymatic activities in rat liver microsome.
Impact of chrysosplenetin on the pharmacokinetics and anti-malarial efficacy of artemisinin against Plasmodium berghei as well as in vitro CYP450 enzymatic activities in rat liver microsome.
Malaria journal (2015-11-06)
Shijie Wei, Hongyan Ji, Bei Yang, Liping Ma, Zhuchun Bei, Xiang Li, Hongwan Dang, Xiaoying Yang, Cheng Liu, Xiuli Wu, Jing Chen
ZUSAMMENFASSUNG
Artemisinin (ART) is an efficacious and safe anti-malarial drugs but has low oral bioavailability and auto-induction profiles during multiple dosing. The pharmacokinetic disadvantages have been found to partially depend on the induction of cytochrome P-450 enzymes by ART and resulted in the therapeutic failure due to insufficient drug levels. The present study, therefore, investigated the impacts of chrysosplenetin (CHR), a polymethoxylated flavonoid from Artemisia annua, on the pharmacokinetics and the anti-malarial efficacy of ART against Plasmodium berghei. The inhibition of CHR on enzymatic activity of CYP1A2, CYP2A, CYP2C19, CYP2D6, CYP2E1, and CYP3A in rat liver microsome was also investigated. IC50, Km, Ki, and inhibitory type of CHR were respectively calculated. Twenty rats were randomly divided into four groups and received three-day oral doses of ART in absence or presence of CHR (in ratio of 1:0, 1:1, 1:2, and 1:4, respectively). Plasma samples were separately harvested for ART pharmacokinetics analysis using a valid liquid chromatography tandem mass spectrometric (LC-MS/MS) method. Female Kunming mice were inoculated by P. berghei K173 strain and pre-exposed to three-day oral administration of ART with or without CHR as pharmacokinetics protocol. Giemsa staining method was applied to calculate percent parasitaemia (%) and inhibition (%). In vitro rat liver microsomal model was employed to elucidate the inhibitory effect of CHR on CYP1A2, CYP2A, CYP2C19, CYP2D6, CYP2E1, and CYP3A. The AUC0-t, Cmax, and t 1/2 of ART increased significantly (P < 0.05 or P < 0.01) as well as declined CLz (P < 0.05 or P < 0.01) after three-day oral doses of ART in presence of CHR (1:2) when compared with ART alone. Also, parasitaemia (%) remarkably attenuated 1.59 folds with 1.63-fold augmented inhibition (%) when the ratio between ART and CHR reached 1:2. CHR itself had no anti-malarial efficacy (P > 0.05). CHR inhibited in vitro activity of CYP1A2 and CYP2C19 (P < 0.01, IC50 = 4.61 and 6.23 μM) in a concentration-response manner. The inhibition did not emerge on CYP2E1 and CYP3A until the CHR concentration exceeded 4.0 μM (P < 0.01, IC50 = 28.17 and 3.38 µM). CHR has no impact on CYP 2A and CYP2D6 (P > 0.05). The inhibition types of CHR on CYP1A2 and CYP3A belonged to noncompetitive and uncompetitive, respectively. Co-administration of ART with CHR in ratio of 1:2 achieved a synergic anti-malarial effect partly because of the noncompetitive or uncompetitive inhibition of CHR of drug-metabolism enzymes, especially CYP3A which is closely related to the auto-induction of ART.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Triethylamin, ≥99%
Sigma-Aldrich
Triethylamin, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Triethylamin, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Natriumphosphat, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Natriumphosphat, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Natriumphosphat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Natriumphosphat monobasisch -Lösung, BioUltra, 5 M in H2O
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumphosphat, BioXtra, ≥99.0%
Sigma-Aldrich
Triethylamin, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Natriumphosphat, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Artemisinin, 98%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Triethylamin, ampule, ≥99.5% (GC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)