Merck
  • Differential expression of cell cycle regulators in CDK5-dependent medullary thyroid carcinoma tumorigenesis.
Differential expression of cell cycle regulators in CDK5-dependent medullary thyroid carcinoma tumorigenesis.
Oncotarget (2015-04-23)
Karine Pozo, Antje Hillmann, Alexander Augustyn, Florian Plattner, Tao Hai, Tanvir Singh, Saleh Ramezani, Xiankai Sun, Roswitha Pfragner, John D Minna, Gilbert J Cote, Herbert Chen, James A Bibb, Fiemu E Nwariaku
ZUSAMMENFASSUNG
Medullary thyroid carcinoma (MTC) is a neuroendocrine cancer of thyroid C-cells, for which few treatment options are available. We have recently reported a role for cyclin-dependent kinase 5 (CDK5) in MTC pathogenesis. We have generated a mouse model, in which MTC proliferation is induced upon conditional overexpression of the CDK5 activator, p25, in C-cells, and arrested by interrupting p25 overexpression. Here, we identify genes and proteins that are differentially expressed in proliferating versus arrested benign mouse MTC. We find that downstream target genes of the tumor suppressor, retinoblastoma protein, including genes encoding cell cycle regulators such as CDKs, cyclins and CDK inhibitors, are significantly upregulated in malignant mouse tumors in a CDK5-dependent manner. Reducing CDK5 activity in human MTC cells down-regulated these cell cycle regulators suggesting that CDK5 activity is critical for cell cycle progression and MTC proliferation. Finally, the same set of cell cycle proteins was consistently overexpressed in human sporadic MTC but not in hereditary MTC. Together these findings suggest that aberrant CDK5 activity precedes cell cycle initiation and thus may function as a tumor-promoting factor facilitating cell cycle protein expression in MTC. Targeting aberrant CDK5 or its downstream effectors may be a strategy to halt MTC tumorigenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-GAPDH-Antikörper, Maus monoklonal in Maus hergestellte Antikörper, clone GAPDH-71.1, purified from hybridoma cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Tantal(V)-ethoxid, 99.98% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Desoxyribonuklease I Rind, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Desoxyribonuklease I Rind, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Tantal(V)-ethoxid, packaged for use in deposition systems
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-phospho-Tau (pThr205) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.