Merck
  • Stability of a highly concentrated solution of epirubicin for conventional transcatheter arterial chemoembolization.
Stability of a highly concentrated solution of epirubicin for conventional transcatheter arterial chemoembolization.
International journal of pharmaceutics (2015-10-11)
Youssef Bennis, Amandine Savry, Florian Correard, Marc Montana, Christophe Sauzet, Laurence Gauthier-Villano, Pascale Pisano, Bertrand Pourroy
ZUSAMMENFASSUNG
Epirubicin is widely used for conventional transcatheter arterial chemoembolization (cTACE) in patients with hepatocellular-carcinoma. However, there is no data about its stability in solution at concentration higher than 2 mg/L, yet needed when mixing it with a standard volume of Lipiodol(®) to produce an efficient water-in-oil emulsion. The aim of this study was therefore to evaluate the stability of a highly concentrated solution of epirubicin for cTACE and verify whether epirubicin solution could be prepared in advance. Fifty milligrams of epirubicin were dissolved in 6 mL of 0.9% sodium chloride and conditioned in brown polypropylene syringe. Physical and chemical stability assays including particles and HPLC-DAD analysis were performed in triplicate, using series of 5 syringes stored over 72 h at 4±2 °C followed by 4h at 22±4°C. Neither weight loss nor pH or spectrum change occurred. No haze or turbidity was observed and the number of subvisible particles was below the recommended limits. Epirubicin concentration remained above 95% of the initial value over the 72 h of storage at +4 °C followed by 4h at 22±4 °C and no degradation was observed. Epirubicin at 50mg/6 mL in 0.9% NaCl conditioned in brown propylene syringe is stable for at least 72 h at 4±2 °C with additional 4h at 22±4 °C allowing its preparation in advance for programmed cTACE and the standardization of its use in clinical practice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Iodwasserstoffsäure, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Iodwasserstoffsäure, contains no stabilizer, ACS reagent, 55%
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Iodwasserstoffsäure, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Methyl-4-hydroxybenzoat, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Iodwasserstoffsäure, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O