Merck
  • Emergence of an Ancestral Glycoprotein Hormone in the Pituitary of the Sea Lamprey, a Basal Vertebrate.
Emergence of an Ancestral Glycoprotein Hormone in the Pituitary of the Sea Lamprey, a Basal Vertebrate.
Endocrinology (2015-06-13)
Stacia A Sower, Wayne A Decatur, Krist N Hausken, Timothy J Marquis, Shannon L Barton, James Gargan, Mihael Freamat, Michael Wilmot, Lian Hollander, Jeffrey A Hall, Masumi Nozaki, Michal Shpilman, Berta Levavi-Sivan
ZUSAMMENFASSUNG
The gnathostome (jawed vertebrates) classical pituitary glycoprotein hormones, FSH, LH, and TSH, consist of a common α-subunit (GpA1) and unique β-subunits (Gpβ1, -2, and -3), whereas a recently identified pituitary glycoprotein hormone, thyrostimulin, consists of GpA2 and GpB5. This paper reports the identification, expression, and function of an ancestral, nonclassical, pituitary heterodimeric glycoprotein hormone (GpH) consisting of the thyrostimulin A2 subunit with the classical β-subunit in the sea lamprey, Petromyzon marinus, a jawless basal vertebrate. Lamprey (l) GpA2, and lGpHβ were shown to form a heterodimer by coimmunoprecipitation of lGpA2 with FLAG-tagged lGpHβ after the overexpression in transiently transfected COS7 cells using a bipromoter vector. Dual-label fluorescent in situ hybridization and immunohistochemistry showed the coexpression of individual subunits in the proximal pars distalis of the pituitary. GnRH-III (1μΜ) significantly increased the expression of lGpHβ and lGpA2 in in vitro pituitary culture. Recombinant lamprey GpH was constructed by tethering the N terminal of lGpA2 to the C terminal of lGpHβ with a linker region composed of six histidine residues followed by three glycine-serine repeats. This recombinant lamprey GpH activated the lamprey glycoprotein hormone receptor I as measured by increased cAMP/luciferase activity. These data are the first to demonstrate a functional, unique glycoprotein heterodimer that is not found in any other vertebrate. These data suggest an intermediate stage of the structure-function of the gonadotropin/thyroid-stimulating hormone in a basal vertebrate, leading to the emergence of the highly specialized gonadotropin hormones and thyroid stimulating hormones in gnathostomes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
o-Toluidin, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Tetrazoliumblauchlorid, suitable for microbiology, ≥90% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Tetrazoliumblauchlorid, used in colorimetric determination of reducing compounds
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Influenza-Hämagglutinin(HA)-Peptid, ≥97% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Argon, ≥99.998%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
o-Toluidin, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Preise und Verfügbarkeit sind derzeit nicht verfügbar.