Merck
  • Influences of exogenous probiotics and tea polyphenols on the production of three acids during the simulated colonic fermentation of maize resistant starch.
Influences of exogenous probiotics and tea polyphenols on the production of three acids during the simulated colonic fermentation of maize resistant starch.
Journal of food science and technology (2015-09-08)
Qian Geng, Xin-Huai Zhao
ZUSAMMENFASSUNG
In the present study, influences of nine probiotics and tea polyphenols on the production of acetic, butyric, and lactic acid from simulated colonic fermentation of maize resistant starch (RS) were investigated. RS was fermented by fecal extracts of healthy adults at 12 g L(-1) and 37 °C for 18-48 h in the presence and absence of exogenous probiotics and tea polyphenols. The added probiotics increased acetic and butyric acid production by 25-216 %. Eubacterium faecalis, Lactobacillus acidophilus, L. casei, and L. helveticus increased lactic acid production by 7-58 %; however, other probiotics decreased lactic acid production. Tea polyphenols facilitated lactic acid production but inhibited acetic and butyric acid production clearly. More importantly, the added probiotics weakened the inhibitory effects of tea polyphenols on the two acids, enhancing acetic and butyric acid production by 152-641 and 825-1,777 %, respectively. It is concluded that both probiotics and tea polyphenols have different impacts on the production of acetic, butyric and lactic acid during the colonic fermentation of RS. The impacts of other probiotics and food components on the colonic fermentation of RS and other dietary fibers should be investigated in future study to clarify their possible interactions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Milchsäure, meets USP testing specifications
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Hexan, anhydrous, 95%
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Milchsäure, natural, ≥85%
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Milchsäure -Lösung, ACS reagent, ≥85%
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR