Merck
  • Characterization of Anti-Citrinin Specific ScFvs Selected from Non-Immunized Mouse Splenocytes by Eukaryotic Ribosome Display.
Characterization of Anti-Citrinin Specific ScFvs Selected from Non-Immunized Mouse Splenocytes by Eukaryotic Ribosome Display.
PloS one (2015-07-02)
Haiwei Cheng, Yifei Chen, Yi Yang, Xueqiu Chen, Xiaolu Guo, Aifang Du
ZUSAMMENFASSUNG
Single chain variable fragments (scFvs) against citrinin (CIT) were selected from a scFv library constructed from the splenocytes of non-immunized mice by an improved eukaryotic ribosome display technology in this study. Bovine serum albumin (BSA)/ CIT-BSA and ovalbumin (OVA)/ CIT-OVA were used as the antigens to select specific anti-CIT scFvs. Eukaryotic in situ RT-PCR method was used to recover the selected mRNA after every affinity selection. After six rounds of ribosome display, expression vector pTIG-TRX carrying specific scFv DNAs were constructed and transformed into Escherichia coli BL21 (DE3) for protein expression. Thirteen positive clones were selected out of which three (designated 23, 68 and 109) showed high binding activity and specificity to CIT by indirect ELISA, while no clone showed binding activity with carrier proteins. The three scFvs showed high specificity to CIT and the cross reactivity with other mycotoxins was below 0.01% as determined by indirect competitive ELISA. These specific scFvs offer a potential novel immunoassay method for CIT residues. This study confirmed the effectiveness of the improved eukaryotic ribosome display system and could be used as a reference for the selection of scFvs specific to other small molecules using ribosome display.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimid -hydrochlorid, commercial grade, powder
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimid -hydrochlorid, ≥99.0% (AT)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimid -hydrochlorid, crystalline
Sigma-Aldrich
Aflatoxin B1, from Aspergillus flavus
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimid -hydrochlorid, BioXtra
Sigma-Aldrich
Fumonisin B1 aus Fusarium moniliforme, ≥98% (HPLC)
Sigma-Aldrich
MES -Lösung, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Citrinin, from Penicillium citrinum, ≥98% (HPLC)
Sigma-Aldrich
4-Chlor-1-Naphthol, crystalline
Sigma-Aldrich
L-Thyroxin Natriumsalz Pentahydrat, ≥98% (HPLC), powder
Sigma-Aldrich
4-Chlor-1-Naphthol, tablet
Sigma-Aldrich
L-Thyroxin Natriumsalz Pentahydrat, γ-irradiated, powder, BioXtra, suitable for cell culture