Merck
  • Heat induces adenosine triphosphate release from mast cells in vitro: a putative mechanism for moxibustion.
Heat induces adenosine triphosphate release from mast cells in vitro: a putative mechanism for moxibustion.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan (2015-08-05)
Lei Hu, Lina Wang, Jianzi Wei, Grygorczyk Ryszard, Xueyong Shen, Schwarz Wolfgang
ZUSAMMENFASSUNG
To investigate the role of adenosine tri-phosphate (ATP) purinergic signaling in mast cells (MCs) modulated by heat to further understand the molecular mechanisms of moxibustion. Skin temperatures induced by monkshood cake moxibustion were evaluated by measuring the Neiguan acupoint (PC 6) from 31 participants with a digital thermocouple thermometer. Temperatures of 43 °C and 52 °C were applied to cultured human leukemia mast cell line HMC-1 in vitro. Calcium fluorescence was applied to detect intracellular Ca2+ ([Ca2+]). Extracellular ATP contents were measured by luciferin-luciferase assay. Maximum skin temperatures mostly ranged from 40-45 °C , but some reached up to 50 °C. Both 43 °C and 52 °C induced MC degranulation, which was accompanied by an increase in [Ca2+] and ATP release. Complexing extracellular Ca2+ with 5 mM ethylene glycol-bis (β-aminoethyl ether)-N,N,N',N'-tetraacetic acid (EGTA) inhibited the noxious, heat-induced elevation of [Ca2+]i and prevented the enhanced ATP secretion by those. cells at 52 °C, but not 43 °C. Monkshood cake moxibustion can generate heat sufficient to trigger cellular events of MCs, including degranulation, [Ca2+]i elevation, and ATP release, suggesting that purinergic signals originating from MCs are possibly the initiating response of acupoints to moxibustion.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
N-Ethylmaleimid, crystalline, ≥98% (HPLC)
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
5-Aminolevulinsäure -hydrochlorid, ≥98%
Sigma-Aldrich
N-Ethylmaleimid, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
5-Aminolevulinsäure -hydrochlorid, BioReagent, suitable for cell culture, powder, ≥98%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
N-Ethylmaleimid, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)