Merck
  • Lapatinib enhances trastuzumab-mediated antibody-dependent cellular cytotoxicity via upregulation of HER2 in malignant mesothelioma cells.
Lapatinib enhances trastuzumab-mediated antibody-dependent cellular cytotoxicity via upregulation of HER2 in malignant mesothelioma cells.
Oncology reports (2015-10-28)
Riki Okita, Katsuhiko Shimizu, Yuji Nojima, Takuro Yukawa, Ai Maeda, Shinsuke Saisho, Masao Nakata
ZUSAMMENFASSUNG
EGFR/HER2 are frequently expressed in MPM tissues, however, no studies have shown the clinical benefit of using EGFR/HER2-targeting drugs in patients with malignant pleural mesothelioma (MPM). It was reported that the tyrosine kinase inhibitor (TKI) lapatinib enhanced trastuzumab-mediated antibody-dependent cellular cytotoxicity (ADCC) in HER2-positive breast cancer, suggesting that this combination is a promising strategy for MPM treatment. The aim of the present study was to explore the possibility of a TKI combined with trastuzumab to enhance ADCC in MPM cells. Five MPM cell lines were used to test the effects of TKIs targeting EGFR (gefitinib, afatinib and lapatinib) on cell proliferation and the expression of the HER family receptor. The combined effects of TKI with trastuzumab on ADCC were evaluated using the LDH release assay. Additionally, MPM cells were isolated from patients and evaluated for lapatinib-induced upregulation of HER family receptors and trastuzumab- or cetuximab‑mediated ADCC. In MPM cell lines, HER2 expression was upregulated by lapatinib, downregulated by afatinib and unaffected by gefitinib. As expected, more trastuzumab bound to MPM cells pretreated with lapatinib than untreated cells, resulting in the enhancement of trastuzumab-mediated ADCC in MPM cells. In patient-derived MPM cells, both HER2 and EGFR were upregulated by lapatinib, resulting in the enhancement of both trastuzumab- and cetuximab-mediated ADCC. Of the three TKIs, only lapatinib enhanced trastuzumab-mediated ADCC via the upregulation of HER2 expression in MPM cells, suggesting that sequential combination of lapatinib and trastuzumab may be a promising strategy for MPM treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
L-Glutamin
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Aphidicolin, ≥98% (HPLC), powder
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
MES -Lösung, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture