Merck
Feasibility study for intraepidermal delivery of proteins using a solid microneedle array.
International journal of pharmaceutics (2015-03-31)
Madeleine Witting, Katja Obst, Markus Pietzsch, Wolfgang Friess, Sarah Hedtrich
ZUSAMMENFASSUNG
Solid microneedles (MN) are a promising tool for dermal drug delivery. Particular focus lies on the field of vaccination due to pain-free, safe, hygienic and patient compliant antigen deposition. Diverse coating techniques and formulations have been developed to preserve vaccine activity and to enable targeted drug deposition in the skin. Process and long-term storage stability of coated MN, however, have not yet been studied in detail. Hence, a feasibility study was conducted determining the appropriate needle length (300 μm) for local intraepidermal protein delivery. Moreover, a protein-stabilizing coating formulation was developed. Coating of the MN resulted in protein concentrations between 10 and 23 μg, 90% of the bioactivity of the model protein asparaginase was maintained for 3 months. Skin experiments verified the intraepidermal deposition of 68.0 ± 11.7% of the coated model protein after single application. Slightly increased interleukin 8 levels right after MN insertion indicated minor skin irritation due to the mechanical piercing stress. Thus, specifically highlighting protein stabilization during storage, we demonstrated that selective intraepidermal deposition of proteins or peptides' using solid MN is a feasible approach.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Methylenblau, certified by the BSC
Sigma-Aldrich
Methylenblau -Lösung, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Rhodamin-B-isothiocyanat, mixed isomers
Sigma-Aldrich
Rhodamin-B-Isothiocyanat, BioReagent, mixed isomers, suitable for protein labeling
Sigma-Aldrich
Methylenblau -Lösung, 0.05 wt. % in H2O
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Methylenblaulösung 1,4 % (w/v) 95 % Ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis