Merck
  • Novel enzymatic synthesis of spacer-linked P(k) trisaccharide targeting for neutralization of Shiga toxin.
Novel enzymatic synthesis of spacer-linked P(k) trisaccharide targeting for neutralization of Shiga toxin.
Journal of biotechnology (2015-06-24)
Tatsuya Kato, Takahiro Oizumi, Makoto Ogata, Akiko Murakawa, Taichi Usui, Enoch Y Park
ZUSAMMENFASSUNG
A novel alkyl spacer-conjugated derivative of P(k) trisaccharide (P(k)), one of the active receptors of Shiga toxins (Stxs; Stx1 and Stx2) produced by pathogenic Escherichia coli (STEC), was designed and synthesized by a combination of cellulase-mediated condensation from Trichoderma reesei and α1,4-galactosyltransferase (LgtC) from Neisseria gonorrhoeae. The specific activity of N. gonorrhoeae LgtC was 66U/mg, which was 13-fold higher than that from N. meningitidis expressed in E. coli. 5-trifluoroacetamidopentyl-β-P(k) (TFAP-P(k)) was synthesized (yield of 86%, based on the amount of TFAP-lactose added) and its binding to Stx1a-B and Stx2a-B was evaluated. The dissociation constants (KDs) of Stx1a-B and Stx2a-B to the spacer-linked P(k), immobilized on a CM5 sensor chip, were 6.8×10(-6) M (kon=4.1×10(1)M(-1)S(-1), koff=2.8×10(-4)S(-1)) and 2.2×10(-5)M (kon=3.9×10(2)M(-1)S(-1), koff=8.6×10(-3)S(-1)), respectively. This result suggests that the monovalent P(k)-derivative, conjugated to a pentylamino group, represents a promising Stx-neutralizing agent. This cellulase-mediated condensation using cellulase and glycosyltransferase is a valuable tool for the synthesis of spacer-linked oligosaccharide.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis