Merck
Reduction of aflatoxins (B₁, B₂, G₁, and G₂) in soybean-based model systems.
Food chemistry (2015-07-21)
Jongin Lee, Jae-Young Her, Kwang-Geun Lee
ZUSAMMENFASSUNG
The effects of chemical, physical, and cooking treatments on the reduction of aflatoxin B1 (AFB1), B2, G1, and G2 in soybean matrix were investigated. A HPLC-FLD with a Kobra cell system was used for the quantitative analysis of aflatoxins (AFs). To decrease the level of AFs during the soaking process, the contaminated soybeans were submerged in organic acid solutions. The reduction rates of AFB1 in 1.0N citric acid, lactic acid, succinic acid, and tartaric acid for 18h were 94.1%, 92.7%, 62.0%, and 95.1%, respectively. In the case of pH and autoclave treatment, the level of AFB1 was significantly decreased during autoclaving process at pH 7.4, 9.0, and 11.1, compared with the non-autoclaved samples (p<0.05). In the case of physical treatment, the heating process at 100 and 150°C for 90min significantly decreased the level of AFB1 by 41.9% and 81.2%, respectively (p<0.05). The reduction rate of AFB1 after cooking was 97.9% for soybean milk and 33.6% for steamed soybeans.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Milchsäure, meets USP testing specifications
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
Milchsäure, natural, ≥85%
Sigma-Aldrich
Milchsäure -Lösung, ACS reagent, ≥85%
Sigma-Aldrich
Bernsteinsäure, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Bernsteinsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Zitronensäure, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Zitronensäure, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Bernsteinsäure, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
2-Hydroxybuttersäure, 97%
Sigma-Aldrich
DL-Milchsäure, 85 % (w/w), syrup
Sigma-Aldrich
Bernsteinsäure, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.