Merck
  • Clean-up of cereal extracts for gas chromatography-tandem quadrupole mass spectrometry pesticide residues analysis using primary secondary amine and C18.
Clean-up of cereal extracts for gas chromatography-tandem quadrupole mass spectrometry pesticide residues analysis using primary secondary amine and C18.
Journal of chromatography. A (2015-11-18)
Susan S Herrmann, Mette E Poulsen
ZUSAMMENFASSUNG
The level of co-extracted matrix in wheat and oat extracts obtained by the QuEChERS method (EN 15662) is high and the occurrence of free fatty acids generates a major matrix peak in TIC chromatograms (rt. 13-22min). Matrix can compromise the analytical performance in pesticide analysis using GC-MS/MS. In order to reduce the amount and the effects of matrix we tested the effect of using six different amounts of primary secondary amine (PSA) (0, 25, 50, 100, 150 and 200mg/ml extract) with and without the addition of six different amounts of C18 (0, 25, 50, 100, 150 and 200mg/ml extract) in the dispersive solid phase extraction (dSPE) procedure. dSPE clean-up using 25mg/ml extract significantly reduced the major matrix peak observed for wheat extracts. Higher amounts of PSA reduced the analytical response for iprodione and malathion. For oat extract 50-150mg PSA/ml extract was needed to obtain equally low intensity of the matrix peak. For oat the analytical responses of the target pesticides generally increased with increasing amount of PSA. C18 had no significant effect on the intensity of the major matrix peaks and even resulted in lower analytical responses for several of the target pesticides. Based on the present study it is concluded that the optimal dSPE clean-up procedure employs 25mg PSA/ml extract for wheat and 150mg PSA/ml extract for oat.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ammoniumacetat, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG