Merck
  • Penetratin-Mediated Transepithelial Insulin Permeation: Importance of Cationic Residues and pH for Complexation and Permeation.
Penetratin-Mediated Transepithelial Insulin Permeation: Importance of Cationic Residues and pH for Complexation and Permeation.
The AAPS journal (2015-05-21)
Mie Kristensen, Henrik Franzyk, Mia Thorne Klausen, Anne Iversen, Jesper Søborg Bahnsen, Rikke Bjerring Skyggebjerg, Vito Foderà, Hanne Mørck Nielsen
ZUSAMMENFASSUNG
Penetratin is a widely used carrier peptide showing promising potential for mucosal delivery of therapeutic proteins. In the present study, the importance of specific penetratin residues and pH was investigated with respect to complexation with insulin and subsequent transepithelial insulin permeation. Besides penetratin, three analogues were studied. The carrier peptide-insulin complexes were characterized in terms of size and morphology at pH 5, 6.5, and 7.4 by dynamic light scattering (DLS) and transmission electron microscopy (TEM), respectively. At pH 7.4 mainly very large complexes were present, while much smaller complexes dominated at pH 5. Presence of arginine residues in the carrier peptide proved to be a prerequisite for complexation with insulin as well as for enhanced transepithelial insulin permeation in vitro. Rearrangement of tryptophan residues resulted in significantly increased insulin permeation as compared to that of the parent penetratin. In general, pre-complexation with penetratin and its analogues at pH 5 gave rise to increased insulin permeation as compared to that observed at pH 7.4; this finding was further supported by a preliminary in vivo study using the parent penetratin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Phenazin-Methosulfat
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
MES -Lösung, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%