Merck
  • Optimization of soybean processing into kinema, a Bacillus-fermented alkaline food, with respect to a minimum level of antinutrients.
Optimization of soybean processing into kinema, a Bacillus-fermented alkaline food, with respect to a minimum level of antinutrients.
Journal of applied microbiology (2015-04-18)
A Sharma, S Kumari, P Wongputtisin, M J R Nout, P K Sarkar
ZUSAMMENFASSUNG
Optimization of traditional processing of soybeans using response surface methodology (RSM) to achieve a minimum level of antinutritional factors (ANFs) in kinema. Central composite rotatable designs were used to optimize the processing stages of kinema preparation. In each stage, the linear or quadratic effects of independent variables were significant in minimizing ANF levels. The predicted optimum condition for soaking was when the raw beans-water ratio was 1 : 10, and the soaking temperature, time and pH were 10°C, 20 h and 8·0 respectively. Here, tannins content (TC), phytic acid content (PAC) and trypsin inhibitor activity (TIA) decreased (P < 0·05). While haemagglutinating activity (HA) level remained unchanged (P < 0·05), total biogenic amines content (TBAC) increased. The optimum condition for cooking was optimally soaked beans-water ratio of 1 : 5, and cooking pressure and time were 1·10 kg cm(-2) and 20 min respectively. Here, TC, PAC, TIA and HA decreased (P < 0·05), but TBAC remained unchanged compared to optimally soaked beans. TC and HA went below the level of detection. The optimum condition for fermentation was obtained when inoculum load was 10(3) total cells g(-1) grits, and fermentation temperature and time were 37°C and 48 h respectively. Fermentation of optimally cooked beans caused a reduction (P < 0·05) of PAC. While TIA remained unchanged (P < 0·05), TBAC increased. In kinema, TC, PAC, TIA and HA decreased (P < 0·05) over raw beans by 100, 61, 71 and 100% respectively. Good agreement was observed between predicted values and experimental values. The processing treatments significantly minimized the level of ANFs in soybeans. RSM was successfully deployed to obtain the optimum condition for kinema-making with a minimum level of ANFs without impairing sensory attributes of the product. The results are useful for commercial production of kinema.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Histamin -dihydrochlorid, ≥99% (TLC), powder
Sigma-Aldrich
4-Aminoantipyrin, reagent grade
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis