Merck
  • Flavonoid profile, antioxidant and cytotoxic activity of different extracts from Algerian Rhamnus alaternus L. bark.
Flavonoid profile, antioxidant and cytotoxic activity of different extracts from Algerian Rhamnus alaternus L. bark.
Pharmacognosy magazine (2015-06-26)
Soulef Boussahel, Antonio Speciale, Saliha Dahamna, Yacine Amar, Irene Bonaccorsi, Francesco Cacciola, Francesco Cimino, Paola Donato, Guido Ferlazzo, Daoud Harzallah, Mariateresa Cristani
ZUSAMMENFASSUNG
Rhamnus alaternus (Rhamnaceae) L. has been traditionally used for treatment of many diseases. In this study, we determined the antioxidant/free radical scavenger properties, the flavonoid profile and the cytotoxicity of aqueous and methanolic extracts obtained by maceration from Algerian R. alaternus bark, like also of aqueous extract prepared by decoction according to the traditional method. This to estimate the usefulness of the drug traditional preparation and compare it with those made in the laboratory. The antioxidant activity of the extracts was evaluated using five different redox-based assays, all involving one redox reaction with the oxidant. High-performance liquid chromatography/diode array detection/electrospray ionization mass spectrometry analysis was used to identify and quantify the flavonoids content. Cytotoxicity on human monocytic leukemia cells (U937) was also carried out. All the extracts tested showed a good antioxidant/free radical scavenger activity and a similar flavonoid fingerprint. However, the methanolic one presented the best antioxidant activity that can be due to the highest flavonoid amount and significantly reduced the proliferation of leukemia cells. The results confirm that the extract prepared by decoction contains efficient antioxidant compounds and this justifies in part the therapeutic and preventive usefulness. Moreover, the methanolic extract exerted excellent cytotoxicity on U937 that could be attributed to kaempferol and rhamnocitrin glycosides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Natriumnitrit, 99.999% trace metals basis
Sigma-Aldrich
Aluminiumchlorid, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Kaliumpersulfat, 99.99% trace metals basis
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Aluminiumchlorid, 99.99% trace metals basis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Aluminiumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, natural, 97%, FG