Merck
  • Melissa officinalis L. decoctions as functional beverages: a bioactive approach and chemical characterization.
Melissa officinalis L. decoctions as functional beverages: a bioactive approach and chemical characterization.
Food & function (2015-06-16)
Márcio Carocho, Lillian Barros, Ricardo C Calhelha, Ana Ćirić, Marina Soković, Celestino Santos-Buelga, Patricia Morales, Isabel C F R Ferreira
ZUSAMMENFASSUNG
Lemon balm (Melissa officinalis L.) is a member of the Lamiaceae family with a long story of human consumption. It has been consumed for decades, directly in food and as a decoction or an infusion for its medicinal purposes. In this manuscript, a detailed chemical characterization of the decoction of this plant is described, encompassing antimicrobial, antioxidant and antitumor activities. Rosmarinic acid and lithospermic acid A were the most abundant phenolic compounds. Quinic acid, fructose, glucose and γ-tocopherol were the most abundant within their groups of molecules. M. officinalis decoctions were active against a wide range of microorganisms, Pseudomonas aeruginosa and Salmonella typhimurium, and Penicillium funiculosum being the most sensitive bacteria and fungi, respectively. The growth inhibition of different human tumor cell lines (mainly MCF-7 and HepG2) was also observed, as also high free radical scavenging activity and reducing power. This manuscript highlights some beneficial effects of these functional beverages.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Hexan, anhydrous, 95%
Sigma-Aldrich
Saccharose, ACS reagent
SAFC
L-Glutamin
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture