Merck
  • Effervescence and graphitized multi-walled carbon nanotubes assisted microextraction for natural antioxidants by ultra high performance liquid chromatography with electrochemical detection and quadrupole time-of-flight tandem mass spectrometry.
Effervescence and graphitized multi-walled carbon nanotubes assisted microextraction for natural antioxidants by ultra high performance liquid chromatography with electrochemical detection and quadrupole time-of-flight tandem mass spectrometry.
Journal of chromatography. A (2015-10-06)
Shu-Ling Wang, Xiao-Qing Pang, Jun Cao, Wan Cao, Jing-Jing Xu, Qiong-Yao Zhu, Qian-Yun Zhang, Li-Qing Peng
ZUSAMMENFASSUNG
In this article, effervescence and graphitized multi-walled carbon nanotubes assisted microextraction was first developed for the extraction of antioxidants in hawthorn samples. The use of an effervescent tablet composed of sodium dihydrogen phosphate, sodium carbonate and micro-scale carboxyl graphitized multi-walled carbon nanotubes (extraction sorbent) was the core of the method. In this study, ultra high performance liquid chromatography coupled with electrochemical detection and quadrupole time-of-flight tandem mass spectrometry was performed for qualitative and quantitative analyses of target analytes in hawthorn foodstuffs. Several experimental factors, such as amount of effervescent salts, the sorbent, elution time and elution solvent, were systematically assessed. Under the optimized conditions, a good linearity with R values better than 0.9980 was obtained. The detection limits estimated at a signal-to-noise ratio of 3:1 were ranging from 0.01 to 0.18ng/mL. These results suggested that the proposed method could be an alternative and promising sample preparation tool in future food analysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Natriumphosphat, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Natriumphosphat, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME-Faseranordnung Polydimethylsiloxan (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub