Merck
Direct writing electrodes using a ball pen for paper-based point-of-care testing.
The Analyst (2015-06-17)
Zedong Li, Fei Li, Jie Hu, Wei Hong Wee, Yu Long Han, Belinda Pingguan-Murphy, Tian Jian Lu, Feng Xu
ZUSAMMENFASSUNG
The integration of paper with an electrochemical device has attracted growing attention for point-of-care testing, where it is of great importance to fabricate electrodes on paper in a low-cost, easy and versatile way. In this work, we report a simple strategy for directly writing electrodes on paper using a pressure-assisted ball pen to form a paper-based electrochemical device (PED). This method is demonstrated to be capable of fabricating electrodes on paper with good electrical conductivity and electrochemical performance, holding great potential to be employed in point-of-care applications, such as in human health diagnostics and food safety detection. As examples, the PEDs fabricated using the developed method are applied for detection of glucose in artificial urine and melamine in sample solutions. Furthermore, our developed strategy is also extended to fabricate PEDs with multi-electrode arrays and write electrodes on non-planar surfaces (e.g., paper cup, human skin), indicating the potential application of our method in other fields, such as fabricating biosensors, paper electronics etc.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Melamin, 99%
Sigma-Aldrich
Natriumphosphat, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Harnsäure, ≥99%, crystalline
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumphosphat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumphosphat, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Natriumphosphat, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Natriumphosphat, BioXtra, ≥99.0%
Sigma-Aldrich
Natriumphosphat, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumphosphat, zweibasisch -Lösung, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Ferrocenmethanol, 97%
Sigma-Aldrich
Natriumphosphat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Natriumphosphat monobasisch -Lösung, BioUltra, 5 M in H2O