Merck
Design of a mediated enzymatic fuel cell to generate power from renewable fuel sources.
Environmental technology (2015-06-24)
Seyda Korkut, Muhammet Samet Kilic
ZUSAMMENFASSUNG
The present work reported a compartment-less enzymatic fuel cell (EFC) based on newly synthesized Poly(pyrrole-2-carboxylic acid-co-3-thiophene acetic acid) film containing glucose oxidase and laccase effectively wired by p-benzoquinone incorporated into the copolymer structure. The resulting system generated a power density of 18.8 µW/cm(2) with 30 mM of glucose addition at +0.94 V at room temperature. Improvements to maximize the power output were ensured with step-by-step optimization of electrode fabrication design and operational parameters for operating the system with renewable fuel sources. We demonstrated that the improved fuel cell could easily harvest glucose produced during photosynthesis to produce electrical energy in a simple, renewable and sustainable way by generating a power density of 10 nW/cm(2) in the plant leaf within 2 min. An EFC for the first time was successfully operated in municipal wastewater which contained glycolytic substances to generate electrical energy with a power output of 3.3 µW/cm(2).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Silbernitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥98%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Silbernitrat, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumcitrat, dreibasisch Dihydrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Natriumcitrat-Dihydrat, ≥99%, FG
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Citronensäure Monohydrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Silbernitrat, 99.9999% trace metals basis
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%