Merck
  • Functional Analysis of Two l-Arabinose Transporters from Filamentous Fungi Reveals Promising Characteristics for Improved Pentose Utilization in Saccharomyces cerevisiae.
Functional Analysis of Two l-Arabinose Transporters from Filamentous Fungi Reveals Promising Characteristics for Improved Pentose Utilization in Saccharomyces cerevisiae.
Applied and environmental microbiology (2015-04-05)
Jingen Li, Jing Xu, Pengli Cai, Bang Wang, Yanhe Ma, J Philipp Benz, Chaoguang Tian
ZUSAMMENFASSUNG
Limited uptake is one of the bottlenecks for l-arabinose fermentation from lignocellulosic hydrolysates in engineered Saccharomyces cerevisiae. This study characterized two novel l-arabinose transporters, LAT-1 from Neurospora crassa and MtLAT-1 from Myceliophthora thermophila. Although the two proteins share high identity (about 83%), they display different substrate specificities. Sugar transport assays using the S. cerevisiae strain EBY.VW4000 indicated that LAT-1 accepts a broad substrate spectrum. In contrast, MtLAT-1 appeared much more specific for l-arabinose. Determination of the kinetic properties of both transporters revealed that the Km values of LAT-1 and MtLAT-1 for l-arabinose were 58.12 ± 4.06 mM and 29.39 ± 3.60 mM, respectively, with corresponding Vmax values of 116.7 ± 3.0 mmol/h/g dry cell weight (DCW) and 10.29 ± 0.35 mmol/h/g DCW, respectively. In addition, both transporters were found to use a proton-coupled symport mechanism and showed only partial inhibition by d-glucose during l-arabinose uptake. Moreover, LAT-1 and MtLAT-1 were expressed in the S. cerevisiae strain BSW2AP containing an l-arabinose metabolic pathway. Both recombinant strains exhibited much faster l-arabinose utilization, greater biomass accumulation, and higher ethanol production than the control strain. In conclusion, because of higher maximum velocities and reduced inhibition by d-glucose, the genes for the two characterized transporters are promising targets for improved l-arabinose utilization and fermentation in S. cerevisiae.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ACS reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Carbonylcyanid-3-chlorphenylhydrazon, ≥97% (TLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.