Merck
  • Schisandrin B induces an Nrf2-mediated thioredoxin expression and suppresses the activation of inflammasome in vitro and in vivo.
Schisandrin B induces an Nrf2-mediated thioredoxin expression and suppresses the activation of inflammasome in vitro and in vivo.
BioFactors (Oxford, England) (2015-08-27)
Pou Kuan Leong, Kam Ming Ko
ZUSAMMENFASSUNG
Reactive oxygen species (ROS)-mediated activation of inflammasome is involved in the development of a wide spectrum of diseases. We aimed to investigate whether (-)schisandrin B [(-)Sch B], a phytochemical that can induce cellular antioxidant response, can suppress the inflammasome activation. Results showed that (-)Sch B can induce an nuclear factor erythroid 2-related factor 2-driven thioredoxin expression in primary peritoneal macrophages and cultured RAW264.7 macrophages. A 4-h priming of peritoneal macrophages with LPS followed by a 30-min incubation with ATP caused the activation of caspase 1 and the release of IL-1β, indicative of inflammasome activation. Although LPS/ATP did not activate inflammasome in RAW264.7 macrophages, it caused the ROS-dependent c-Jun N-terminal kinase1/2 (JNK1/2) activation and an associated lactate dehydrogenase (LDH) release in RAW264.7 macrophages, an indication of cytotoxicity. (-)Sch B suppressed the LPS/ATP-induced activation of caspase 1 and release of IL-1β in peritoneal macrophages. (-)Sch B also attenuated the LPS/ATP-induced ROS production, JNK1/2 activation and LDH release in RAW264.7 macrophages. The ability of (-)Sch B to suppress LPS/ATP-mediated inflammation in vitro was further confirmed by an animal study, in which schisandrin B treatment (2 mmol/kg p.o.) ameliorated the Imject Alum-induced peritonitis, as indicated by suppressions of caspase1 activation and plasma IL-1β level. The ensemble of results suggests that (-)Sch B may offer a promising prospect for preventing the inflammasome-mediated disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, ACS reagent, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, BioReagent, suitable for insect cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, tablet
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aluminium-Kaliumsulfat Dodecahydrat, BioXtra, ≥98.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.