Merck
  • Novel synthetic pyridyl analogues of CDDO-Imidazolide are useful new tools in cancer prevention.
Novel synthetic pyridyl analogues of CDDO-Imidazolide are useful new tools in cancer prevention.
Pharmacological research (2015-08-05)
Martine Cao, Evans O Onyango, Charlotte R Williams, Darlene B Royce, Gordon W Gribble, Michael B Sporn, Karen T Liby
ZUSAMMENFASSUNG
Two new analogues of CDDO-Imidazolide (CDDO-Im), namely 1-[2-Cyano-3,12-dioxooleana-1,9(11)-dien-28-oyl]-4(-pyridin-2-yl)-1H-imidazole ("CDDO-2P-Im") and 1-[2-Cyano-3,12-dioxooleana-1,9(11)-dien-28-oyl]-4(-pyridin-3-yl)-1H-imidazole ("CDDO-3P-Im") have been synthesized and tested for their potential use as chemopreventive drugs. At nanomolar concentrations, they were equipotent to CDDO-Im for inducing differentiation and apoptosis in U937 leukemia cells. As inflammation and oxidative stress contribute to carcinogenesis, we also assessed their cytoprotective potential. The new compounds suppressed inducible nitric oxide synthase (iNOS) expression in RAW264.7 macrophage-like cells and significantly elevated heme oxygenase-1 (HO-1) and quinone reductase (NQO1) mRNA and protein levels in various mouse tissues in vivo. Most importantly, pharmacokinetic studies performed in vitro in human plasma and in vivo showed that each new analogue was more stable than CDDO-Im. Much higher concentrations of the new derivatives were found in mouse liver, lung, pancreas and kidney after gavage in contrast to CDDO-Im. Because of their better bioavailability and their excellent anti-inflammatory profile in vitro, CDDO-2P-Im and CDDO-3P-Im were tested for prevention in a highly relevant mouse lung cancer model, in which A/J mice develop lung carcinomas after injection of vinyl carbamate, a potent carcinogen. CDDO-2P-Im and CDDO-3P-Im were as effective as CDDO-Im for reducing the size and the severity of the lung tumors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
p-Benzochinon, reagent grade, ≥98%
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%