Merck
  • Structural and functional screening in human induced-pluripotent stem cell-derived cardiomyocytes accurately identifies cardiotoxicity of multiple drug types.
Structural and functional screening in human induced-pluripotent stem cell-derived cardiomyocytes accurately identifies cardiotoxicity of multiple drug types.
Toxicology and applied pharmacology (2015-04-07)
Kimberly R Doherty, Dominique R Talbert, Patricia B Trusk, Diarmuid M Moran, Scott A Shell, Sarah Bacus
ZUSAMMENFASSUNG
Safety pharmacology studies that evaluate new drug entities for potential cardiac liability remain a critical component of drug development. Current studies have shown that in vitro tests utilizing human induced pluripotent stem cell-derived cardiomyocytes (hiPS-CM) may be beneficial for preclinical risk evaluation. We recently demonstrated that an in vitro multi-parameter test panel assessing overall cardiac health and function could accurately reflect the associated clinical cardiotoxicity of 4 FDA-approved targeted oncology agents using hiPS-CM. The present studies expand upon this initial observation to assess whether this in vitro screen could detect cardiotoxicity across multiple drug classes with known clinical cardiac risks. Thus, 24 drugs were examined for their effect on both structural (viability, reactive oxygen species generation, lipid formation, troponin secretion) and functional (beating activity) endpoints in hiPS-CM. Using this screen, the cardiac-safe drugs showed no effects on any of the tests in our panel. However, 16 of 18 compounds with known clinical cardiac risk showed drug-induced changes in hiPS-CM by at least one method. Moreover, when taking into account the Cmax values, these 16 compounds could be further classified depending on whether the effects were structural, functional, or both. Overall, the most sensitive test assessed cardiac beating using the xCELLigence platform (88.9%) while the structural endpoints provided additional insight into the mechanism of cardiotoxicity for several drugs. These studies show that a multi-parameter approach examining both cardiac cell health and function in hiPS-CM provides a comprehensive and robust assessment that can aid in the determination of potential cardiac liability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cytosin, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Terfenadin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Astemizol, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Captopril, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®
Preise und Verfügbarkeit sind derzeit nicht verfügbar.