Merck
Construction of RNA-Quantum Dot Chimera for Nanoscale Resistive Biomemory Application.
ACS nano (2015-07-03)
Taek Lee, Ajay Kumar Yagati, Fengmei Pi, Ashwani Sharma, Jeong-Woo Choi, Peixuan Guo
ZUSAMMENFASSUNG
RNA nanotechnology offers advantages to construct thermally and chemically stable nanoparticles with well-defined shape and structure. Here we report the development of an RNA-QD (quantum dot) chimera for resistive biomolecular memory application. Each QD holds two copies of the pRNA three-way junction (pRNA-3WJ) of the bacteriophage phi29 DNA packaging motor. The fixed quantity of two RNAs per QD was achieved by immobilizing the pRNA-3WJ with a Sephadex aptamer for resin binding. Two thiolated pRNA-3WJ serve as two feet of the chimera that stand on the gold plate. The RNA nanostructure served as both an insulator and a mediator to provide defined distance between the QD and gold. Immobilization of the chimera nanoparticle was confirmed with scanning tunneling microscopy. As revealed by scanning tunneling spectroscopy, the conjugated pRNA-3WJ-QD chimera exhibited an excellent electrical bistability signal for biomolecular memory function, demonstrating great potential for the development of resistive biomolecular memory and a nano-bio-inspired electronic device for information processing and computing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diethylpyrocarbonat, 96% (NT)
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Borsäure, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Ethidiumbromid, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Diethylpyrocarbonat, 96% (NT)
Sigma-Aldrich
2-Methyl-2-propanthiol, 99%
Sigma-Aldrich
Diethylpyrocarbonat, 96% (NT)
Sigma-Aldrich
Borsäure, 99.999% trace metals basis
Sigma-Aldrich
Borsäure, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Ethidiumbromid, ~95% (HPLC)
Sigma-Aldrich
Borsäure, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Ethidiumbromid -Lösung, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Borsäure, 99.97% trace metals basis
Sigma-Aldrich
Borsäure-11B, ≥99 atom % 11B
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Borsäure, BioXtra, ≥99.5%
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Borsäure, tablet, 1 g boric acid per tablet