Merck
  • Inhibition of glycogen synthase kinase-3β prevents sympathetic hyperinnervation in infarcted rats.
Inhibition of glycogen synthase kinase-3β prevents sympathetic hyperinnervation in infarcted rats.
Experimental biology and medicine (Maywood, N.J.) (2015-01-13)
Tsung-Ming Lee, Shinn-Zong Lin, Nen-Chung Chang
ZUSAMMENFASSUNG
We have demonstrated that nerve growth factor (NGF) expression in the myocardium is selectively increased during chronic stage of myocardial infarction, resulting in sympathetic hyperinnervation. Glycogen synthase kinase-3 (GSK-3) signal has been shown to play key roles in the regulation of cytoskeletal assembly during axon regeneration. We assessed whether lithium, a GSK-3 inhibitor, attenuates cardiac sympathetic reinnervation after myocardial infarction through attenuated NGF expression and Tau expression. Twenty-four hours after ligation of the anterior descending artery, male Wistar rats were randomized to either LiCl or SB216763, chemically unrelated inhibitors of GSK-3β, a combination of LiCl and SB216763, or vehicle for four weeks. Myocardial norepinephrine levels revealed a significant elevation in vehicle-treated rats compared with sham-operated rats, consistent with excessive sympathetic reinnervation after infarction. Immunohistochemical analysis for sympathetic nerve also confirmed the change of myocardial norepinephrine. This was paralleled by a significant upregulation of NGF protein and mRNA in the vehicle-treated rats, which was reduced after administering either LiCl, SB216763, or combination. Arrhythmic scores during programmed stimulation in the vehicle-treated rats were significantly higher than those treated with GSK-3 inhibitors. Addition of SB216763 did not have additional beneficial effects compared with those seen in rats treated with LiCl alone. Furthermore, lithium treatment increased Tau1 and decreased AT8 and AT180 levels. Chronic use of lithium after infarction, resulting in attenuated sympathetic reinnervation by GSK-3 inhibition, may modify the arrhythmogenic response to programmed electrical stimulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Lithiumchlorid -Lösung, 8 M, Molecular Biology, ≥99%
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Anilin, ACS reagent, ≥99.5%
Sigma-Aldrich
Lithiumchlorid, Molecular Biology, ≥99%
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Lithiumchlorid, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent