Merck
  • Dysregulation of Th1, Th2, Th17, and T regulatory cell-related transcription factor signaling in children with autism.
Dysregulation of Th1, Th2, Th17, and T regulatory cell-related transcription factor signaling in children with autism.
Molecular neurobiology (2016-06-28)
Sheikh Fayaz Ahmad, Khairy M A Zoheir, Mushtaq Ahmad Ansari, Ahmed Nadeem, Saleh A Bakheet, Laila Yousef Al-Ayadhi, Mohammad Zeed Alzahrani, Othman A Al-Shabanah, Mohammed M Al-Harbi, Sabry M Attia
ZUSAMMENFASSUNG
Autism is a neurodevelopmental disorder characterized by stereotypic repetitive behaviors, impaired social interactions, and communication deficits. Numerous immune system abnormalities have been described in individuals with autism including abnormalities in the ratio of Th1/Th2/Th17 cells; however, the expression of the transcription factors responsible for the regulation and differentiation of Th1/Th2/Th17/Treg cells has not previously been evaluated. Peripheral blood mononuclear cells (PBMCs) from children with autism (AU) or typically developing (TD) control children were stimulated with phorbol-12-myristate 13-acetate (PMA) and ionomycin in the presence of brefeldin A. The expressions of Foxp3, RORγt, STAT-3, T-bet, and GATA-3 mRNAs and proteins were then assessed. Our study shows that children with AU displayed altered immune profiles and function, characterized by a systemic deficit of Foxp3
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Heparin Natriumsalz aus Schweinedarm, Grade I-A, ≥180 USP units/mg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.