Merck
Titriplex® solution B for the determination of alkaline earth metals in water 1 ml 10 mg CaO/l using 100 ml of water, Titripur®
Titriplex® solution B
Titriplex® solution B for the determination of alkaline earth metals in water 1 ml 10 mg CaO/l using 100 ml of water, Titripur®
Products
Categoria de produto
Marca
Forma física
Linha de produto