Merck
  • Hepatocyte-specific loss of GPS2 in mice reduces non-alcoholic steatohepatitis via activation of PPARα.
Hepatocyte-specific loss of GPS2 in mice reduces non-alcoholic steatohepatitis via activation of PPARα.
Nature communications (2019-04-13)
Ning Liang, Anastasius Damdimopoulos, Saioa Goñi, Zhiqiang Huang, Lise-Lotte Vedin, Tomas Jakobsson, Marco Giudici, Osman Ahmed, Matteo Pedrelli, Serena Barilla, Fawaz Alzaid, Arturo Mendoza, Tarja Schröder, Raoul Kuiper, Paolo Parini, Anthony Hollenberg, Philippe Lefebvre, Sven Francque, Luc Van Gaal, Bart Staels, Nicolas Venteclef, Eckardt Treuter, Rongrong Fan
RESUMO
Obesity triggers the development of non-alcoholic fatty liver disease (NAFLD), which involves alterations of regulatory transcription networks and epigenomes in hepatocytes. Here we demonstrate that G protein pathway suppressor 2 (GPS2), a subunit of the nuclear receptor corepressor (NCOR) and histone deacetylase 3 (HDAC3) complex, has a central role in these alterations and accelerates the progression of NAFLD towards non-alcoholic steatohepatitis (NASH). Hepatocyte-specific Gps2 knockout in mice alleviates the development of diet-induced steatosis and fibrosis and causes activation of lipid catabolic genes. Integrative cistrome, epigenome and transcriptome analysis identifies the lipid-sensing peroxisome proliferator-activated receptor α (PPARα, NR1C1) as a direct GPS2 target. Liver gene expression data from human patients reveal that Gps2 expression positively correlates with a NASH/fibrosis gene signature. Collectively, our data suggest that the GPS2-PPARα partnership in hepatocytes coordinates the progression of NAFLD in mice and in humans and thus might be of therapeutic interest.
MATERIAIS
Número do produto
Marca
Descrição do produto
Millipore
ANTI-FLAG®, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-PPAR α Antibody, ascites fluid, Chemicon®