Merck
Isolation of microprotoplasts from a partially synchronized suspension culture of Citrus unshiu.
Journal of plant physiology (2006-10-13)
Qinghua Zhang, Jihong Liu, Xiuxin Deng
RESUMO
The effect of hydroxyurea (HU) and amiprophos-methyl (APM) on the synchronization of suspension cultures of Satsuma mandarin (Citrus unshiu) and micronucleation of the suspension cells sequentially treated with both, HU and APM, were investigated. When suspension cultures in early-log phase were treated with 4 or 10mM HU for 24h, the number of cells in the S-phase and the mitotic index (MI) increased significantly. Exposure of the early-log phase suspension culture to 32microM APM led to a marked increase in the MI 12 and 24h after treatment, while higher as well as lower concentrations (16, 24 and 48microM) had no effect. Suspension cultures subjected to sequential treatment, e.g. pretreatment with 10mM HU for 24h followed by treatment with 32microM APM for 24h, also showed a considerably increased MI. Furthermore, 61.5% of the protoplasts isolated from the sequentially treated suspension cells were micronucleated, whereas only 3.6% of the control protoplasts, isolated from untreated cells, showed micronucleation. Ultra-centrifugation of the micronucleated protoplasts generated microprotoplasts of different sizes, most of them below 5microM in diameter, with 1 or few chromosomes. The potential application of microprotoplasts in citrus genetic improvement is discussed.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Amiprofos methyl, PESTANAL®, analytical standard