- Isolated lingual dystonia : responding to anticholinergic treatment.
Isolated lingual dystonia : responding to anticholinergic treatment.
The Journal of the Association of Physicians of India (2011-09-06)
Archana Verma, Ramakant Yadav
PMID21887917
RESUMO
We report a 42 year-old woman with 6 month history of isolated lingual dystonia induced during speaking and was treated with anticholinergic drug. She experienced a marked improvement in her abnormal tongue movements. Primary lingual dystonia is a rare presentation.