Merck
An RNA biding protein, Y14 interacts with and modulates STAT3 activation.
Biochemical and biophysical research communications (2008-05-28)
Norihiko Ohbayashi, Naohisa Taira, Shiho Kawakami, Sumihito Togi, Noriko Sato, Osamu Ikeda, Shinya Kamitani, Ryuta Muromoto, Yuichi Sekine, Tadashi Matsuda
RESUMO
Signal transducer and activator of transcription 3 (STAT3), which mediates biological actions in many physiological processes, is activated by cytokines and growth factors via specific tyrosine-phosphorylation, dimerization, and nuclear translocation. To clarify the molecular mechanisms underlying the regulation of STAT3 activation, we performed yeast two-hybrid screening. We identified Y14, an RNA-binding protein, as a novel STAT3 binding partner. Y14 bound to STAT3 through the C-terminal region of STAT3 in vivo. Importantly, small-interfering RNA-mediated reduction of endogenous Y14 expression decreased IL-6-induced tyrosine-phosphorylation, nuclear accumulation, and DNA-binding activity of STAT3, as well as IL-6/STAT3-dependent gene expression. These results indicate that Y14 interacts with STAT3 and regulates the transcriptional activation of STAT3 by influencing the tyrosine-phosphorylation of STAT3.