- CDX2 upregulates SLC26A3 gene expression in intestinal epithelial cells.
CDX2 upregulates SLC26A3 gene expression in intestinal epithelial cells.
American journal of physiology. Gastrointestinal and liver physiology (2017-06-03)
Ishita Chatterjee, Anoop Kumar, Rosa María Castilla-Madrigal, Oscar Pellon-Cardenas, Ravinder K Gill, Waddah A Alrefai, Alip Borthakur, Michael Verzi, Pradeep K Dudeja
PMID28572085
RESUMO
SLC26A3 [downregulated in adenoma (DRA)] plays a key role in mammalian intestinal NaCl absorption, in that it mediates apical membrane Cl
MATERIAIS
Número do produto
Marca
Descrição do produto