Merck
  • Overexpression of KIFC1 and its association with spheroid formation in esophageal squamous cell carcinoma.
Overexpression of KIFC1 and its association with spheroid formation in esophageal squamous cell carcinoma.
Pathology, research and practice (2017-10-02)
Takeharu Imai, Naohide Oue, Yuji Yamamoto, Ryuichi Asai, Naohiro Uraoka, Kazuhiro Sentani, Kazuhiro Yoshida, Wataru Yasui
RESUMO
Esophageal squamous cell carcinoma (ESCC) is one of the most common human cancers. We previously reported that KIFC1 is involved in gastric cancer pathogenesis and that KIFC1 plays an important role in gastric cancer spheroid colony formation. However, the significance of KIFC1 in ESCC has not been examined. In the present study, we analyzed the expression and distribution of KIFC1 in 132 ESCC cases by immunohistochemistry. In contrast to weak or no staining of KIFC1 in non-neoplastic mucosa, ESCC tissue showed stronger, more extensive KIFC1 staining. In total, 95 (72%) of 132 ESCC cases were positive for KIFC1. Immunostaining of ALDH1 was also performed, and KIFC1-positive ESCC cases were significantly frequently found in ALDH1-positive ESCC cases compared with ALDH1-negative ESCC cases. Spheroid colony formation is an effective method to characterize CSCs, thus we analyzed sphere number and size at 15days in ESCC cells downregulated for KIFC1 by siRNA transfection. Both the number and size of sphere from TE-1 cells were significantly reduced in KIFC1 siRNA-transfected TE-1 cells than in negative control siRNA-transfected cells. These results suggest that KIFC1 plays an important role in ESCC pathogenesis.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
MISSION® esiRNA, targeting human KIFC1