- Polymeric ionic liquid-assembled graphene-immobilized silica composite for selective isolation of human serum albumin from human whole blood.
Polymeric ionic liquid-assembled graphene-immobilized silica composite for selective isolation of human serum albumin from human whole blood.
Analytical and bioanalytical chemistry (2017-12-01)
Jiawei Liu, Yixun Liang, Jiwei Shen, Quan Bai
PMID29184996
RESUMO
Polymeric ionic liquids (PILs) with 1-vinyl-3-ethylimidazolium cations and two different anions of Br