MilliporeSigma
  • Microcystin-LR altered mRNA and protein expression of endoplasmic reticulum stress signaling molecules related to hepatic lipid metabolism abnormalities in mice.
Microcystin-LR altered mRNA and protein expression of endoplasmic reticulum stress signaling molecules related to hepatic lipid metabolism abnormalities in mice.
Environmental toxicology and pharmacology (2015-06-22)
Wendi Qin, Xuxiang Zhang, Liuyan Yang, Lizhi Xu, Zongyao Zhang, Jun Wu, Yaping Wang
ABSTRACT
To explore the effects of microcystin-LR (MC-LR), a hepatotoxin, on the incidence of liver lipid metabolism abnormality, and the potential molecular mechanisms of action, healthy male Balb/c mice were intraperitoneally injected with MC-LR at doses of 0, 5, 10, and 20 μg/kg/d for 14 days. Hepatic histopathology and serum lipid parameters of mice were determined, and the changes of mRNA and protein expression of endoplasmic reticulum (ER) stress signaling molecules related to the lipid metabolism abnormalities in the livers of mice were investigated by quantitative real-time polymerase chain reaction (qPCR) and Western blotting, respectively. The results indicated that 5-20 μg/kg/d MC-LR altered serum lipid parameters and caused hepatic steatosis. MC-LR treatment at 10 or 20 μg/kg/d changed mRNA and protein expression of ER stress signaling molecules, including upregulation of mRNA and protein expression of activating transcription factor 6 (ATF6), pancreatic ER eukaryotic translation initiation factor 2α (eIF-2α) kinase (PERK), and eIF-2α. MC-LR exposure at 10 or 20 μg/kg/d also altered mRNA and protein expression of downstream factors and genes of ER stress signaling pathways, including the downregulation of sterol regulatory element binding protein 1c (SREBP-1c) and fatty acid synthase (FASn), and upregulation of acetyl-coenzyme A carboxylase α (ACACA) and glycogen synthase kinase 3β (Gsk-3β). Our results reveal that ER stress plays a significant role in hepatic lipid metabolism abnormalities in mice exposed to MC-LR.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%