Merck
Oral L-carnitine: metabolite formation and hemodialysis.
Current drug metabolism (2006-11-01)
Marcus A Bain, Randall Faull, Robert W Milne, Allan M Evans
ZUSAMMENFASSUNG
L-Carnitine has important roles in intermediary metabolism and patients with end-stage renal disease who are undergoing hemodialysis may develop a secondary L-carnitine deficiency. The extent of accumulation of the metabolites trimethylamine and trimethylamine-N-oxide when L-carnitine is administered orally has not been investigated previously in this population. Oral L-carnitine at a dose of 1 g daily was administered for twelve days to six patients with end-stage renal disease undergoing hemodialysis thrice weekly. Pre-dialysis plasma concentrations of L-carnitine (mean +/- SD) increased significantly (P < 0.05) from day 1 (baseline; 32.4 +/- 6.1 microM) to day 8 (66.1 +/- 13.8 microM) remaining constant thereafter. Although plasma levels of trimethylamine remained unaltered, the pre-dialysis plasma concentrations of trimethylamine-N-oxide increased significantly (P < 0.05) from day 1 (289.1 +/- 236.1 microM) to day 12 (529.0 +/- 237.9 microM). The hemodialysis clearances for L-carnitine, trimethylamine and trimethylamine-N-oxide were 14.3 +/- 8.2, 14.1 +/- 10.6 and 12.4 +/- 5.4 L/h, respectively, indicating their efficient removal by dialysis. Oral administration of L-carnitine at a dose of 1 g daily increases plasma concentrations of this substance to physiological levels in patients with end-stage renal disease who are undergoing hemodialysis. However, concerns about the possible deleterious consequences of such a dosage regimen still remain given that plasma concentrations of trimethylamine-N-oxide were continually rising and approximately doubled in a two-week period.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Aceton, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Aceton, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Zink, dust, <10 μm, ≥98%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Zink, granular, 20-30 mesh, ACS reagent, ≥99.8%
Supelco
Aceton, analytical standard
Sigma-Aldrich
Zink, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Inosin, ≥99% (HPLC)
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trimethylamin-N-oxid, 95%
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Cadaverin, 95%
Sigma-Aldrich
Dimethylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Phenylessigsäure, 99%
Sigma-Aldrich
Dimethylamin, anhydrous, ≥99%
Sigma-Aldrich
Aceton, histological grade, ≥99.5%