Merck
  • VhuD facilitates electron flow from H2 or formate to heterodisulfide reductase in Methanococcus maripaludis.
VhuD facilitates electron flow from H2 or formate to heterodisulfide reductase in Methanococcus maripaludis.
Journal of bacteriology (2013-09-17)
Kyle C Costa, Thomas J Lie, Qin Xia, John A Leigh
ZUSAMMENFASSUNG
Flavin-based electron bifurcation has recently been characterized as an essential energy conservation mechanism that is utilized by hydrogenotrophic methanogenic Archaea to generate low-potential electrons in an ATP-independent manner. Electron bifurcation likely takes place at the flavin associated with the α subunit of heterodisulfide reductase (HdrA). In Methanococcus maripaludis the electrons for this reaction come from either formate or H2 via formate dehydrogenase (Fdh) or Hdr-associated hydrogenase (Vhu). However, how these enzymes bind to HdrA to deliver electrons is unknown. Here, we present evidence that the δ subunit of hydrogenase (VhuD) is central to the interaction of both enzymes with HdrA. When M. maripaludis is grown under conditions where both Fdh and Vhu are expressed, these enzymes compete for binding to VhuD, which in turn binds to HdrA. Under these conditions, both enzymes are fully functional and are bound to VhuD in substoichiometric quantities. We also show that Fdh copurifies specifically with VhuD in the absence of other hydrogenase subunits. Surprisingly, in the absence of Vhu, growth on hydrogen still occurs; we show that this involves F420-reducing hydrogenase. The data presented here represent an initial characterization of specific protein interactions centered on Hdr in a hydrogenotrophic methanogen that utilizes multiple electron donors for growth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Ameisensäure, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Supelco
Ammoniumformat, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Ameisensäure, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Natriumformiat, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Sigma-Aldrich
Ammoniumformat, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Natriumformiat, reagent grade, 97%
Sigma-Aldrich
Kaliumformiat, ReagentPlus®, 99%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Formiatdehydrogenase aus Candida boidinii, lyophilized powder, 5.0-15.0 units/mg protein
Sigma-Aldrich
Natriumformiat, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Calciumformat, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Cäsiumformiat, 98%
Sigma-Aldrich
Natriumformiat, 99.998% trace metals basis
Sigma-Aldrich
Natriumformiat-13C, 99 atom % 13C
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
Supelco
Calciumformat, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Thallium(I)-formiat, 97%
Sigma-Aldrich
Kaliumformiat, BioUltra, ≥99.0% (NT)