Merck
Guided bone regeneration using chitosan-collagen membranes in dog dehiscence-type defect model.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons (2014-01-21)
Xiaojing Li, Xinmu Wang, Tengfei Zhao, Bo Gao, Yuwen Miao, Dandan Zhang, Yan Dong
ZUSAMMENFASSUNG
The purpose of the present study was to compare a newly developed chitosan-collagen membrane (CCM) with a standard collagen membrane (SCM) regarding their effects on guided bone regeneration. The right mandibular premolars and first molar were extracted from 12 beagle dogs. Four months later, acute buccal dehiscence-type defects (4 × 3 mm in height and width) were surgically created after implant site preparation. The defects were randomly assigned to 4 different groups: CCM-1 (weight ratio of chitosan to collagen of 40:1), CCM-2 (weight ratio of chitosan to collagen of 20:1), SCM, and vehicle control. The dogs were sacrificed after 4, 8, and 12 weeks of healing for radiographic examination, histologic observation, and histometric analysis. The membrane-treated sites showed more bone formation than the control sites, although no statistically significant differences were found between the membrane-treated sites and the control sites for new bone-to-implant contact and new bone-filled area at any point. At 8 weeks, the new bone height for the membrane-treated sites was significantly greater statistically than that of the untreated group (P < .05). At 12 weeks, the CCM-1 group showed significantly greater new bone height (1.91 ± 0.25 mm) than the untreated group (1.20 ± 0.34 mm; P < .05). However, the CCMs did not show any statistically significant differences compared with the SCMs for any assessed parameter. The results of the present study have shown that the developed CCMs can enhance bone regeneration and could be a candidate for use in guided bone regeneration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Kollagen, Typ I -Lösung aus Rattenschwanz, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Rattenschwanz, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Kollagen aus Achillessehne vom Rind, powder, suitable for substrate for collagenase
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Kollagen aus chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen Typ IV aus human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Chitosan aus Garnelenschalen, low-viscous
Sigma-Aldrich
Kollagen aus bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen aus bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Kollagen aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Kaninchenhaut, Bornstein and Traub Type I, powder