Merck
Additive effect of repeated corneal collagen cross-linking in keratoconus.
Journal of refractive surgery (Thorofare, N.J. : 1995) (2014-10-08)
Farhad Hafezi, David Tabibian, Olivier Richoz
ZUSAMMENFASSUNG
To report the long-term clinical outcome in a patient diagnosed as having bilateral progressive keratoconus who received a single corneal collagen cross-linking (CXL) treatment in the right eye and repeated CXL in the left eye. Observational case report. Topographical changes were assessed by high-resolution Scheimpflug imaging. The right eye underwent a standard epithelium-off CXL procedure in February 2008, followed by the left eye 4 weeks later. In 2012, the left eye was treated with CXL for a second time. Irradiation was performed in all cases at a fluence of 5.4 J/cm². Energy settings were 30 minutes @ 3 mW/cm² for the CXL procedures performed in 2008, and 10 minutes @ 9 mW/cm² for the second CXL procedure of the left eye that was performed in 2012. The right eye that underwent a single CXL procedure showed a flattening of keratometry values between 2008 and 2012, followed by stabilization. The left eye showed a similar flattening effect between 2008 and 2012, followed by another flattening effect after the second CXL procedure and accompanied by a distinct increase in corrected distance visual acuity. Following repeated CXL, the corneal stroma and endothelium remained inconspicuous, and postoperative haze and visibility of the stromal demarcation line was similar to what is usually observed after a single CXL procedure. Whether the additive flattening effect of the anterior surface observed in this single case goes along with an additive increase in biomechanical stiffness remains to be seen.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen, Typ I -Lösung aus Rattenschwanz, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Rattenschwanz, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Kollagen aus Achillessehne vom Rind, powder, suitable for substrate for collagenase
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Kollagen aus chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Supelco
(−)-Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kollagen Typ IV aus human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
Kollagen aus bovine tracheal cartilage, Bornstein and Traub Type II, powder
USP
Riboflavin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Kollagen aus bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Kollagen aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Riboflavin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Kollagen aus Kaninchenhaut, Bornstein and Traub Type I, powder