Merck
  • Relationship between elasticity and collagen fiber content in breast disease: a preliminary report.
Relationship between elasticity and collagen fiber content in breast disease: a preliminary report.
Ultrasonics (2014-12-04)
Zhi Li Wang, Lu Sun, Ye Li, Nan Li
ZUSAMMENFASSUNG
To investigate the differences in elasticity and collagen fiber content between malignant and benign breast lesions, and to study the relationship between shear wave elasticity and the content of collagen fiber in extracellular matrix (ECM). Between May 2012 to May 2013, 106 patients with 116 breast lesions who were referred to our center for ultrasound-guided biopsy of a sonographically apparent breast lesion underwent shear wave elasticity examination. The specimen underwent Van Gieson (VG) dye and Image-Pro Plus 5.1 software was used to quantitatively analyze area of collagen fiber. Malignant lesions exhibited significantly higher maximum elasticity, mean elasticity, and elasticity ratio between lesions and surrounding parenchyma (140.43±70.16 kPa, 63.11±33.68 kPa, 3.49±1.95) than benign lesions (54.64±48.53 kPa, 34.52±25.23 kPa, 2.25±1.48) (t=5.329, t=4.382, t=4.487, P<0.001). The content of collagen fiber of malignant lesions was significantly higher than that of benign lesions (t=8.437, p=0.000). There was a positive correlation between max elasticity and the content of fiber collagen (r=0.746). The elasticity of breast lesions has a close correlation with the content of collagen fiber, which might have an important impact on tissue stiffness of breast lesions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen, Typ I -Lösung aus Rattenschwanz, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Rattenschwanz, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Achillessehne vom Rind, powder, suitable for substrate for collagenase
Sigma-Aldrich
Kollagen aus chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen Typ IV aus human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Kollagen aus bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen aus bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Kollagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Kollagen aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Kaninchenhaut, Bornstein and Traub Type I, powder