Merck
  • Melatonin promotes hepatic differentiation of human dental pulp stem cells: clinical implications for the prevention of liver fibrosis.
Melatonin promotes hepatic differentiation of human dental pulp stem cells: clinical implications for the prevention of liver fibrosis.
Journal of pineal research (2014-11-29)
Young-Ah Cho, Kwantae Noh, Seong-Suk Jue, So-Youn Lee, Eun-Cheol Kim
ZUSAMMENFASSUNG
Melatonin's effect on hepatic differentiation of stem cells remains unclear. The aim of this study was to investigate the action of melatonin on hepatic differentiation as well as its related signaling pathways of human dental pulp stem cells (hDPSCs) and to examine the therapeutic effects of a combination of melatonin and hDPSC transplantation on carbon tetrachloride (CCl4 )-induced liver fibrosis in mice. In vitro hepatic differentiation was assessed by periodic acid-Schiff (PAS) staining and mRNA expression for hepatocyte markers. Liver fibrosis model was established by injecting 0.5 mL/kg CCl4 followed by treatment with melatonin (5 mg/kg, twice a week) and hDPSCs. In vivo therapeutic effects were evaluated by histopathology and by means of liver function tests including measurement of alanine transaminase (ALT), aspartate transaminase (AST), and ammonia levels. Melatonin promoted hepatic differentiation based on mRNA expression of differentiation markers and PAS-stained glycogen-laden cells. In addition, melatonin increased bone morphogenic protein (BMP)-2 expression and Smad1/5/8 phosphorylation, which was blocked by the BMP antagonist noggin. Furthermore, melatonin activated p38, extracellular signal-regulated kinase (ERK), and nuclear factor-κB (NF-κB) in hDPSCs. Melatonin-induced hepatic differentiation was attenuated by inhibitors of BMP, p38, ERK, and NF-κB. Compared to treatment of CCl4 -injured mice with either melatonin or hDPSC transplantation alone, the combination of melatonin and hDPSC significantly suppressed liver fibrosis and restored ALT, AST, and ammonia levels. For the first time, this study demonstrates that melatonin promotes hepatic differentiation of hDPSCs by modulating the BMP, p38, ERK, and NF-κB pathway. Combined treatment of grafted hDPSCs and melatonin could be a viable approach for the treatment of liver cirrhosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
DL-Alanin, ≥99% (HPLC)
USP
Melatonin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
DL-Alanin, ≥99%, FCC, FG
Alanin, European Pharmacopoeia (EP) Reference Standard
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethason, tested according to Ph. Eur.
Selen, foil, 25x25mm, thickness 3mm, 99.95%
Dexamethason, British Pharmacopoeia (BP) Assay Standard
Dexamethason für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Dexamethason für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Selen, pellets, < 5mm, ≥99.999%