Merck
  • Isopropoxy-carvacrol, a derivative obtained from carvacrol, reduces acute inflammation and nociception in rodents.
Isopropoxy-carvacrol, a derivative obtained from carvacrol, reduces acute inflammation and nociception in rodents.
Basic & clinical pharmacology & toxicology (2014-10-03)
Rangel R Bonfim, Igor O Paiva-Souza, Janaína P Moraes, Denyson S Pereira, Cliomar A Santos, Danielle G Santana, Sara M Thomazzi, Jamile N S Ferro, Emiliano O Barreto, Damião P Sousa, Enilton A Camargo
ZUSAMMENFASSUNG
Monoterpenes, compounds mainly presented in essential oils, have important pharmacological actions. Isopropoxy-carvacrol (IPC) is a derivative of the monoterpene carvacrol, and its pharmacological properties have not yet been investigated. The aim of this study was to analyse the acute anti-inflammatory and antinociceptive properties of IPC. Mice (25–30 g) and rats (150–230 g) were pre-treated (i.p.) with IPC at the doses of 10, 30 or 100 mg/kg or vehicle (Tween 80, 0.5%), 30 min. before injection of the phlogistic agents. Both the first and the second phases of formalin-induced nociception were significantly reduced by IPC (100 mg/kg). Injection of carrageenan in mice paw reduced the threshold of stimulus intensity, applied with an analgesymeter, necessary to cause paw withdrawal, which was significantly reduced by 100 mg/kg of IPC. The area under curve (0–4 hr) of rat paw oedema induced by injection of carrageenan was also significantly diminished by the administration of IPC (100 mg/kg). Administration of 12-O-tetradecanoylphorbol-13-acetate (TPA) markedly increased mice ear oedema and myeloperidase (MPO) activity. Topical co-administration of IPC (0.3–3 mg/ear) during the induction did not affect TPA-induced ear oedema, but significantly decreased MPO activity in the ears, when compared with the vehicle. In in vitro experiments, IPC reduced lipoperoxidation induced by different stimuli, showed nitric oxide scavenger activity and did not interfere with murine macrophage viability in concentrations up to 100 lg/mL. These results demonstrate that IPC exerts acute anti-inflammatory and antinociceptive activities, suggesting that it may represent an alternative in the development of new future therapeutic strategies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
o-Dianisidin, peroxidase substrate
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥96.0% (AT)
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Dexamethason, meets USP testing specifications
USP
Cetrimoniumbromid, United States Pharmacopeia (USP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Supelco
o-Dianisidin, for spectrophotometric det. of Au, NO2-, Ce(IV), for the detection of Au, Co, Cu, SCN-, V, ≥97.0%
Supelco
Hexadecyltrimethylammoniumbromid, analytical standard
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Hexadecyltrimethylammoniumbromid, suitable for ion pair chromatography, LiChropur
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethason, tested according to Ph. Eur.
Dexamethason, British Pharmacopoeia (BP) Assay Standard
Dexamethason für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Dexamethason für die Systemeignung, European Pharmacopoeia (EP) Reference Standard