Merck
  • Quantitation of Albumin in Serum Using "Turn-on" Fluorescent Probe with Aggregation-Enhanced Emission Characteristics.
Quantitation of Albumin in Serum Using "Turn-on" Fluorescent Probe with Aggregation-Enhanced Emission Characteristics.
ACS applied materials & interfaces (2015-11-11)
Wangyang Li, Didi Chen, Huan Wang, Shanshan Luo, Lichao Dong, Yahui Zhang, Jianbing Shi, Bin Tong, Yuping Dong
ZUSAMMENFASSUNG
An aggregation-enhanced emission active luminogen named as sodium 4,4'4″-(3,4-diphenyl-1H-pyrrole-1,2,5-triyl)tribenzoate (DP-TPPNa) with propeller construction was synthesized and developed as a "turn on" fluorescent probe for in situ quantitation of albumin in blood serum. The DP-TPPNa fluorescence intensity was linearly correlated with the concentration of two serum albumins, bovine serum albumin (BSA) and human serum albumin (HSA), in pure PBS buffer in the ranges of 2.18-70 and 1.68-100 μg/mL, respectively. The detection limits were as low as 2.18 μg/mL for BSA and 1.68 μg/mL for HSA. The response time of fluorescence to serum albumin (SA) was very short (below 6 s), which achieved real-time detection. It also showed high selectivity to SA because other components in serum barely interfere with the detection of DP-TPPNa to SA, enabling in situ quantitative detection of SA without isolation from serum. DP-TPPNa was successfully applied for the quantitative detection of BSA in fetal bovine serum. The mechanism of fluorescent turn-on behavior was elucidated utilizing an unfolding process induced by guanidine hydrochloride, which revealed a capture process via selective hydrophobic interaction and hydrogen bonding between luminogen and SA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Natriumhydroxid, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Natriumhydroxid, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natronlauge, purum, ≥32%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Natriumhydroxid, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O