Merck
Atrial stretch delays gastric emptying of liquids in awake rats.
Life sciences (2013-01-29)
R C Palheta, M T B Silva, H L G Barbosa, A D N Pinheiro, K V V Cardoso, J R V Graça, P J C Magalhães, R B Oliveira, A A Santos
ABSTRACT
We previously reported that mechanical atrial stretch (AS) by balloon distention increased gastric tonus in anesthetized rats. The present study evaluated the effect of AS on the gastric emptying of a liquid test meal in awake rats and its underlying neural mechanisms. Anesthetized male rats received a balloon catheter into the right atrium and a gastrostomy cannula. The next day, mean arterial pressure (MAP), heart rate (HR), central venous pressure (CVP), and cardiac output (CO) were continuously monitored. After the first 20min of monitoring (basal interval), the balloon was either distended or not (control) with 30, 50, or 70μl saline for 5min. Fifteen minutes later, the rats received the test meal (glucose solution with phenol red), and fractional gastric dye retention was determined 10, 20, or 30min later. Heart rate and CVP values were transiently increased by 50 or 70μl AS but not 30μl AS, whereas gastric emptying was slower after 30, 50, or 70μl AS than after sham distention. Subdiaphragmatic vagotomy or splanchnicotomy+celiac ganglionectomy and capsaicin, ondansetron, hexamethonium, L-NAME, and glibenclamide treatment prevented the AS-induced delay in gastric emptying, whereas atropine and guanethidine treatment failed to prevent it. Atrial stretch inhibited the gastric emptying of liquid via non-adrenergic and non-cholinergic pathways that activate nitric oxide-K(+)ATP channels.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Glibenclamide, ≥99% (HPLC)
Supelco
Ondansetron Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ondansetron hydrochloride dihydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Hexamethonium bromide
Supelco
Atropine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hexamethonium chloride, solid (hygroscopic)
Sigma-Aldrich
Glibenclamide, meets USP testing specifications
Sigma-Aldrich
Hexamethonium hydroxide solution, ~0.1 M in H2O
Ondansetron hydrochloride dihydrate, European Pharmacopoeia (EP) Reference Standard
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
Ondansetron for LC system suitability, European Pharmacopoeia (EP) Reference Standard
Ondansetron for TLC system suitability, European Pharmacopoeia (EP) Reference Standard
Glibenclamide, European Pharmacopoeia (EP) Reference Standard