Merck
  • Collagen-induced arthritis: severity and immune response attenuation using multivalent N-acetyl glucosamine.
Collagen-induced arthritis: severity and immune response attenuation using multivalent N-acetyl glucosamine.
Clinical and experimental immunology (2014-03-05)
J Richter, K Capková, V Hříbalová, L Vannucci, I Danyi, M Malý, A Fišerová
ABSTRACT
Rheumatoid arthritis is an autoimmunity leading to considerable impairment of quality of life. N-acetyl glucosamine (GlcNAc) has been described previously as a potent modulator of experimental arthritis in animal models and is used for osteoarthritis treatment in humans, praised for its lack of adverse effects. In this study we present a comprehensive immunological analysis of multivalent GlcNAc-terminated glycoconjugate (GC) application in the treatment of collagen-induced arthritis (CIA) and its clinical outcome. We used immunohistochemistry and FACS to describe conditions on the inflammation site. Systemic and clinical effects were evaluated by FACS, cytotoxicity assay, ELISA, cytometric bead array (CBA), RT-PCR and clinical scoring. We found reduced inflammatory infiltration, NKG2D expression on NK and suppression of T, B and antigen-presenting cells (APC) in the synovia. On the systemic level, GCs prevented the activation of monocyte- and B cell-derived APCs, the rise of TNF-α and IFN-γ levels, and subsequent type II collagen (CII)-specific IgG2a formation. Moreover, we detected an increase of anti-inflammatory IL-4 mRNA in the spleen. Similar to the synovia, the GCs caused a significant reduction of NKG2D-expressing NK cells in the spleen without influencing their lytic function. GCs effectively postponed the onset of arthritic symptoms, reduced their severity and in 18% (GN8P) and 31% (GN4C) of the cases completely prevented their appearance. Our data prove that GlcNAc glycoconjugates prevent the inflammatory response, involving proinflammatory cytokine rise, APC activation and NKG2D expression, leading to the attenuation of clinical symptoms. These results support the glycobiological approach to the treatment of collagen-induced arthritis/rheumatoid arthritis (CIA/RA) as a way of bringing new prospects for more effective therapeutic interventions.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acido acetico, glacial, ACS reagent, ≥99.7%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acido acetico, glacial, ReagentPlus®, ≥99%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acido acetico, glacial, ≥99.99% trace metals basis
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acido acetico, suitable for HPLC
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Ioduro di propidio, ≥94.0% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acido acetico, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acido acetico, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
N-acetil-D-glucosammina, ≥99% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
N-acetil-D-glucosammina, suitable for cell culture, BioReagent
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Propidium iodide solution
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Sodium azide, BioXtra
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acido acetico, suitable for luminescence, BioUltra, ≥99.5% (GC)
Prezzi e disponibilità al momento non sono disponibili
USP
Acido acetico, United States Pharmacopeia (USP) Reference Standard
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
N-acetil-D-glucosammina, ≥95% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Supelco
Acido acetico, analytical standard
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Ioduro di propidio, ≥94% (HPLC)
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acido acetico, ≥99.5%, FCC, FG
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acido acetico, natural, ≥99.5%, FG
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acido acetico, glacial, puriss., 99-100%
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Prezzi e disponibilità al momento non sono disponibili
Supelco
N-acetil-D-glucosammina, Pharmaceutical Secondary Standard; Certified Reference Material
Prezzi e disponibilità al momento non sono disponibili
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Prezzi e disponibilità al momento non sono disponibili
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Prezzi e disponibilità al momento non sono disponibili
Millipore
Acido acetico, suitable for microbiology
Prezzi e disponibilità al momento non sono disponibili